KEY240035531 Key Organics / BIONET

2,3-dichloropyridin-4-amine

  • MDL number MFCD01646113
  • Cas Number : 184416-83-9
  • Molecular Weight : 163.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 152 - 153
Iupac Name 2,3-dichloro-4-pyridinamine
Inchi Code 1S/C5H4Cl2N2/c6-4-3(8)1-2-9-5(4)7/h1-2H,(H2,8,9)
Inchi Key DVUKWILERQFZFC-UHFFFAOYSA-N

Description

Melting Point 152 - 153
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100966

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/KD-0223_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart