KEY240035532 Key Organics / BIONET

[4'-(trifluoromethyl)-[1,1'-biphenyl]-4-yl]methanol

  • MDL number MFCD01862515
  • Cas Number : 457889-46-2
  • Molecular Weight : 252.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 147 - 149
Iupac Name [4'-(trifluoromethyl)[1,1'-biphenyl]-4-yl]methanol
Inchi Code 1S/C14H11F3O/c15-14(16,17)13-7-5-12(6-8-13)11-3-1-10(9-18)2-4-11/h1-8,18H,9H2
Inchi Key YCVFDTRUBSFXSA-UHFFFAOYSA-N

Description

Melting Point 147 - 149
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101005

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/KD-0742_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart