KEY240035533 Key Organics / BIONET

ethyl 4-hydroxy-1-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate

  • MDL number MFCD09031522
  • Cas Number : 853105-37-0
  • Molecular Weight : 197.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117 - 119
Purity 95
Storage temp. RT
Physical Form White to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101006

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/KD-0744_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart