KEY240035536 Key Organics / BIONET

5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD03094951
  • Cas Number : 129768-28-1
  • Molecular Weight : 180.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 171 - 173
Iupac Name 5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid
Inchi Code 1S/C5H3F3N2O2/c6-5(7,8)3-1-2(4(11)12)9-10-3/h1H,(H,9,10)(H,11,12)
Inchi Key CIVNBJPTGRMGRS-UHFFFAOYSA-N

Description

Melting Point 171 - 173
Purity 95%
Storage temp. Store under N2 at 4
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100970

Safety Information

signalword Warning
pcodes P411,P422, P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/KD-0237_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart