KEY240035537 Key Organics / BIONET

2-amino-4H,5H,7H-thieno[2,3-c]pyran-3-carboxamide

  • MDL number MFCD12087875
  • Cas Number : 848324-33-4
  • Molecular Weight : 198.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222 - 223
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101011

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/KD-0751_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart