KEY240887135 Key Organics / BIONET

methyl 4,6-dichloro-5-nitropyridine-3-carboxylate

  • MDL number MFCD13193110
  • Cas Number : 89247-05-2
  • Molecular Weight : 251.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65 - 66
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100990

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/KD-0725_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart