KEY240887137 Key Organics / BIONET

3-bromo-5-hydroxy-1,2-dihydroquinolin-2-one

  • MDL number MFCD13193084
  • Cas Number : 1192263-85-6
  • Molecular Weight : 240.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 300
Purity 0.90
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101004

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/KD-0740_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart