KEY240887304 Key Organics / BIONET

propan-2-yl N-[(2S)-7-bromo-1H,2H,3H,4H-cyclopenta[b]indol-2-yl]carbamate

  • MDL number MFCD13193067
  • Cas Number : 1212283-31-2
  • Molecular Weight : 337.217

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174 - 176
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97607

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/H-20253_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart