KEY240887315 Key Organics / BIONET

2,4-dibromo-5-(trifluoromethyl)aniline

  • MDL number MFCD12547852
  • Cas Number : 24115-24-0
  • Molecular Weight : 318.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp LMS
Iupac Name 2,4-dibromo-5-(trifluoromethyl)aniline
Inchi Code 1S/C7H4Br2F3N/c8-4-2-5(9)6(13)1-3(4)7(10,11)12/h1-2H,13H2
Inchi Key LHDNHSKFNNUECM-UHFFFAOYSA-N

Description

Melting Point LMS
Purity 95%
Physical Form Low Melting Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101803

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/LD-0065_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart