KEY240887316 Key Organics / BIONET

5-[4-nitro-3-(trifluoromethyl)phenyl]-2H-1,2,3,4-tetrazole

  • MDL number MFCD13193118
  • Cas Number : 1192263-88-9
  • Molecular Weight : 259.148

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93 - 95
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101806

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/LD-0084_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart