KEY240887333 Key Organics / BIONET

6-bromo-1,2-benzoxazol-3-ol

  • MDL number MFCD13193078
  • Cas Number : 65685-51-0
  • Molecular Weight : 214.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 219 - 221
Iupac Name 6-bromo-1,2-benzisoxazol-3-ol
Inchi Code 1S/C7H4BrNO2/c8-4-1-2-5-6(3-4)11-9-7(5)10/h1-3H,(H,9,10)
Inchi Key ZXDOSRLQEYIVJC-UHFFFAOYSA-N

Description

Melting Point 219 - 221
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101845

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/LD-0722_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart