KEY240887336 Key Organics / BIONET

1H,2H,3H-pyrazolo[3,4-b]pyridin-3-one

  • MDL number MFCD03013435
  • Cas Number : 2942-43-0
  • Molecular Weight : 135.126

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 234 - 236
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100981

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/KD-0714_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart