KEY240887337 Key Organics / BIONET

methyl 1-methyl-1H-1,3-benzodiazole-5-carboxylate

  • MDL number MFCD04492113
  • Cas Number : 131020-36-5
  • Molecular Weight : 190.202

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109 - 111
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100997

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/KD-0733_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart