KEY241404751 Key Organics / BIONET

2-chloro-6-cyclopropylpyridine-3-carbonitrile

  • MDL number MFCD13193091
  • Cas Number : 1198475-35-2
  • Molecular Weight : 178.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100 - 102
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102170

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MC-0779_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart