KEY241404753 Key Organics / BIONET

3-bromo-6-nitroimidazo[1,2-a]pyridine

  • MDL number MFCD13193088
  • Cas Number : 52310-42-6
  • Molecular Weight : 242.032

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140 - 142
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102173

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MC-0786_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart