KEY241404763 Key Organics / BIONET

2H,4H,5H,6H,7H-pyrazolo[4,3-c]pyridine dihydrochloride

  • MDL number MFCD13368261
  • Cas Number : 410544-19-3
  • Molecular Weight : 196.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 260 - 264
Iupac Name 4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine dihydrochloride
Inchi Code 1S/C6H9N3.2ClH/c1-2-7-3-5-4-8-9-6(1)5;;/h4,7H,1-3H2,(H,8,9);2*1H
Inchi Key ZPIFAYFILYYZIG-UHFFFAOYSA-N

Description

Melting Point 260 - 264
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102238

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MD-0732_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart