KEY241404770 Key Organics / BIONET

methyl 5-bromo-2-hydroxy-3-iodobenzoate

  • MDL number MFCD00464941
  • Cas Number : 18071-51-7
  • Molecular Weight : 356.941

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126 - 128
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101793

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/LD-0027_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart