KEY241404772 Key Organics / BIONET

methyl 5-bromo-2-hydroxy-3-nitrobenzoate

  • MDL number MFCD00458681
  • Cas Number : 91983-31-2
  • Molecular Weight : 276.042

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147 - 149
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102177

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MD-0013_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart