KEY241404773 Key Organics / BIONET

6-chloro-7-nitro-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD02660643
  • Cas Number : 116862-22-7
  • Molecular Weight : 228.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 283 - 285
Purity 95
Storage temp. RT
Physical Form Brown to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102229

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/MD-0715_SDS.pdf
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart