KEY241404936 Key Organics / BIONET

7-fluoro-3,4-dihydro-2H-1,4-benzoxazine

  • MDL number MFCD11848177
  • Cas Number : 56346-41-9
  • Molecular Weight : 153.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 41 - 43
Iupac Name 7-fluoro-3,4-dihydro-2H-1,4-benzoxazine
Inchi Code 1S/C8H8FNO/c9-6-1-2-7-8(5-6)11-4-3-10-7/h1-2,5,10H,3-4H2
Inchi Key VRBQMPCMLZNPSK-UHFFFAOYSA-N

Description

Melting Point 41 - 43
Purity 95%
Physical Form Liquid,Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102231

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/MD-0717_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart