KEY241404945 Key Organics / BIONET

2,5-bis(benzyloxy)benzaldehyde

  • MDL number MFCD02380367
  • Cas Number : 6109-54-2
  • Molecular Weight : 318.372

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98 - 100
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102191

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MD-0059_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart