KEY241404963 Key Organics / BIONET

(1E)-1-(dimethylamino)-4-methylpent-1-en-3-one

  • MDL number MFCD09859309
  • Cas Number : 5782-56-9
  • Molecular Weight : 141.214

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90 C @ 1.5mBar
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102219

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MD-0703_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart