KEY241404976 Key Organics / BIONET

4-iodo-2-methoxypyridine-3-carbaldehyde

  • MDL number MFCD05662396
  • Cas Number : 158669-26-2
  • Molecular Weight : 263.034

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96 - 97
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102225

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MD-0710_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart