KEY241404977 Key Organics / BIONET

ethyl 4-chloro-1-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate

  • MDL number MFCD08234670
  • Cas Number : 821791-58-6
  • Molecular Weight : 215.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 94 - 96
Iupac Name ethyl 4-chloro-1-methyl-6-oxo-1,6-dihydro-3-pyridinecarboxylate
Inchi Code 1S/C9H10ClNO3/c1-3-14-9(13)6-5-11(2)8(12)4-7(6)10/h4-5H,3H2,1-2H3
Inchi Key WGSWOEOJYNQPRP-UHFFFAOYSA-N

Description

Melting Point 94 - 96
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101858

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/LD-0740_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart