KEY245858988 Key Organics / BIONET

5-iodo-2-oxo-6-(propan-2-yl)-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD14581669
  • Cas Number : 1203898-26-3
  • Molecular Weight : 288.088

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 245 - 247
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106765

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/ND-0707_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart