KEY245858990 Key Organics / BIONET

4-amino-3-hydroxybenzenesulfonic acid

  • MDL number MFCD14560533
  • Cas Number : 2592-14-5
  • Molecular Weight : 189.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108255

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3052_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart