KEY245858993 Key Organics / BIONET

2-bromo-4-nitro-6-(trifluoromethyl)aniline

  • MDL number MFCD14560537
  • Cas Number : 400-66-8
  • Molecular Weight : 285.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 142 - 143
Iupac Name 2-bromo-4-nitro-6-(trifluoromethyl)aniline
Inchi Code 1S/C7H4BrF3N2O2/c8-5-2-3(13(14)15)1-4(6(5)12)7(9,10)11/h1-2H,12H2
Inchi Key VTSOQYVBDQONBG-UHFFFAOYSA-N

Description

Melting Point 142 - 143
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106743

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/ND-0047_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart