KEY245858997 Key Organics / BIONET

5-chloro-2-nitrobenzenol

  • MDL number MFCD09260849
  • Cas Number : 611-07-4
  • Molecular Weight : 173.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form Yellow to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108291

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-3119_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart