KEY245859050 Key Organics / BIONET

2-{[1-(tert-butoxycarbonyl)-4-piperidinyl]oxy}propanoic acid

  • MDL number MFCD14581674
  • Cas Number : 952486-65-6
  • Molecular Weight : 273.329

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108258

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3056_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart