KEY245859054 Key Organics / BIONET

2-(1H-1,2,4-triazol-1-yl)benzoic acid hydrochloride

  • MDL number MFCD14581678
  • Cas Number : 1203898-11-6
  • Molecular Weight : 225.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108289

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3116_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart