KEY245859068 Key Organics / BIONET

methyl 5-(4-methoxyphenyl)-5-oxopentanoate

  • MDL number MFCD00229176
  • Cas Number : 1847-68-3
  • Molecular Weight : 236.267

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108250

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3047_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart