KEY245859070 Key Organics / BIONET

6-nitro-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD06656903
  • Cas Number : 24623-24-3
  • Molecular Weight : 177.159

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66585

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1Z-5050_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart