KEY245859078 Key Organics / BIONET

6-chloro-9-methyl-9H-purine

  • MDL number MFCD00127840
  • Cas Number : 2346-74-9
  • Molecular Weight : 168.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-9-methyl-9H-purine
Inchi Code 1S/C6H5ClN4/c1-11-3-10-4-5(7)8-2-9-6(4)11/h2-3H,1H3
Inchi Key UEZNSCCMEMUEMO-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108268

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-3075_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart