KEY250699796 Key Organics / BIONET

{4H,5H,6H,7H,8H-pyrazolo[1,5-a][1,4]diazepin-2-yl}methanol

  • MDL number MFCD14584770
  • Cas Number : 1221792-15-9
  • Molecular Weight : 167.212

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94206

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9X-0601_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart