KEY250699807 Key Organics / BIONET

methyl 3-amino-4-(benzyloxy)-5-methoxybenzoate

  • MDL number MFCD14584787
  • Cas Number : 1221791-79-2
  • Molecular Weight : 287.315

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89 - 91
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94632

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0069_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart