KEY250699810 Key Organics / BIONET

3-(benzyloxy)-2-bromo-4,6-diiodopyridine

  • MDL number MFCD14584794
  • Cas Number : 1221792-34-2
  • Molecular Weight : 515.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 122 - 124
Iupac Name 3-(benzyloxy)-2-bromo-4,6-diiodopyridine
Inchi Code 1S/C12H8BrI2NO/c13-12-11(9(14)6-10(15)16-12)17-7-8-4-2-1-3-5-8/h1-6H,7H2
Inchi Key JAWFFRMFLICJDV-UHFFFAOYSA-N

Description

Melting Point 122 - 124
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94640

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AE-0100_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart