KEY250699811 Key Organics / BIONET

1-benzyl-3-(benzyloxy)-5-iodo-2-methyl-1,4-dihydropyridin-4-one

  • MDL number MFCD14584795
  • Cas Number : 1221792-03-5
  • Molecular Weight : 431.273

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154 - 156
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94641

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0104_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart