KEY250699813 Key Organics / BIONET

ethyl 3-cyano-6-phenylpyridine-2-carboxylate

  • MDL number MFCD14584809
  • Cas Number : 1221792-25-1
  • Molecular Weight : 252.273

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99 - 101
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94663

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0701_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart