KEY250699816 Key Organics / BIONET

8-benzyl-1-thia-4,8-diazaspiro[4.5]decane

  • MDL number MFCD14584829
  • Cas Number : 710268-50-1
  • Molecular Weight : 248.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76 - 77
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94692

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0741_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart