KEY250699819 Key Organics / BIONET

methyl 3-amino-5-bromo-2-methoxybenzoate hydrochloride

  • MDL number MFCD14584846
  • Cas Number : 926036-31-9
  • Molecular Weight : 296.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199 - 201 C
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106748

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/ND-0059_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart