KEY250699822 Key Organics / BIONET

5-chloro-2-hydroxy-3-nitrophenyl benzoate

  • MDL number MFCD14584784
  • Cas Number : 194294-25-2
  • Molecular Weight : 293.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95 - 96
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94629

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0059_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart