KEY250699965 Key Organics / BIONET

methyl 7-methoxy-2-methyl-1,3-benzoxazole-5-carboxylate

  • MDL number MFCD14584796
  • Cas Number : 1197944-26-5
  • Molecular Weight : 221.212

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134 - 136
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94642

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0108_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart