KEY250699966 Key Organics / BIONET

2,3,5-trichloropyridine-4-carbonyl chloride

  • MDL number MFCD14584797
  • Cas Number : 1221791-81-6
  • Molecular Weight : 244.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp Oil
Iupac Name 2,3,5-trichloroisonicotinoyl chloride
Inchi Code 1S/C6HCl4NO/c7-2-1-11-5(9)4(8)3(2)6(10)12/h1H
Inchi Key ODLHIOFZIWQERB-UHFFFAOYSA-N

Description

Melting Point Oil
Purity 95%
Storage temp. Store under N2 at -18
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94643

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/AE-0206_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart