KEY250699969 Key Organics / BIONET

2,3,5-trichloropyridine-4-carbonitrile

  • MDL number MFCD14584800
  • Cas Number : 1221791-83-8
  • Molecular Weight : 207.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58 - 59
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94646

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0211_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart