KEY250699970 Key Organics / BIONET

2-amino-3-chloro-5-(trifluoromethyl)pyridine-4-carbonitrile

  • MDL number MFCD14584801
  • Cas Number : 1221572-26-4
  • Molecular Weight : 221.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210 - 211
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94647

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0214_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart