KEY250699977 Key Organics / BIONET

ethyl 3-chloro-4-cyano-5-(trifluoromethyl)pyridine-2-carboxylate

  • MDL number MFCD14584811
  • Cas Number : 1221792-27-3
  • Molecular Weight : 278.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59 - 60
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94665

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0703_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart