KEY250699978 Key Organics / BIONET

3-chloro-4-cyano-5-(trifluoromethyl)pyridine-2-carboxylic acid

  • MDL number MFCD14584813
  • Cas Number : 1221791-55-4
  • Molecular Weight : 250.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122 (dec)
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94667

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0707_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart