KEY250699989 Key Organics / BIONET

3,4-dihydro-2H-1,4-benzothiazine-6-carboxylic acid

  • MDL number MFCD14584825
  • Cas Number : 226259-31-0
  • Molecular Weight : 195.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209 - 210
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94684

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0729_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart