KEY250699994 Key Organics / BIONET

tert-butyl 1-thia-4,8-diazaspiro[4.5]decane-8-carboxylate hydrochloride

  • MDL number MFCD14584832
  • Cas Number : 1221792-10-4
  • Molecular Weight : 294.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197 - 199
Purity 95
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94695

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0744_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart