KEY250699996 Key Organics / BIONET

2-bromo-1-(3-methoxy-4-nitrophenyl)ethan-1-one

  • MDL number MFCD14584834
  • Cas Number : 90725-63-6
  • Molecular Weight : 274.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98 - 100
Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94698

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0748_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart